Tisk…

3,5-Diiodosalicylaldehyde ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,5-Diiodo-2-hydroxybenzaldehyde

L05783.09 L05783.18
L05783.09EA 2110 CZK
L05783.09 L05783.18
3,5-Diiodosalicylaldehyde ≥98%
3,5-Diiodosalicylaldehyde

Purity: 98+% CAS No.: 2631-77-8 Molecular Formula: C7H4I2O2 Formula Weight: 373.92 Melting Point: 106-110° Beilstein Reference: 1102552

Vzorec: C₇H₄I₂O₂
MW: 373,92 g/mol
Bod tání: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003321
CAS číslo: 2631-77-8
EINECS: 220-117-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR