Tisk…

2-Phenoxyaniline ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Amino-2-phenoxybenzol

L05558.14 L05558.22
L05558.14EA 1930 CZK
L05558.14 L05558.22
2-Phenoxyaniline ≥98%
2-Phenoxyaniline

Purity: 99% CAS No.: 2688-84-8 Molecular Formula: C12H11NO Formula Weight: 185.23 Flash Point: 110°(230°F) Melting Point: 46-48° Boiling Point: 170°/18mm Beilstein Reference: 777766 MDL No.: MFCD00035765

Vzorec: C₁₂H₁₁NO
MW: 185,23 g/mol
Bod varu: 170 °C (18 mmHg)
Bod tání: 46…48 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035765
CAS číslo: 2688-84-8
EINECS: 220-254-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR