4-(Methylthio)aniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-(Methylmercapto)aniline
L04950.06
L04950.14
L04950.06EA
1940
CZK
L04950.06
L04950.14
4-(Methylthio)aniline ≥98%
4-(Methylthio)aniline
Purity: 98%
CAS No.: 104-96-1
Molecular Formula: C7H9NS
Formula Weight: 139.23
Flash Point:
Vzorec:
C₇H₉NS Bod varu: 272…273 °C Bod tání: 156…158 °C Hustota: 1,120 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007889 CAS číslo: 104-96-1 EINECS: 203-256-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...