Tisk…

N-(3-Hydroxypropyl)phthalimide ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-(3-Hydroxypropyl)isoindoline-1,3-dione

L04507.09 L04507.18
L04507.09EA 1020 CZK
L04507.09 L04507.18
N-(3-Hydroxypropyl)phthalimide ≥98%
N-(3-Hydroxypropyl)phthalimide

Purity: 98% CAS No.: 883-44-3 Molecular Formula: C11H11NO3 Formula Weight: 205.22 Melting Point: 75-80° Beilstein Reference: 158489

Vzorec: C₁₁H₁₁NO₃
Bod tání: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00023097
CAS číslo: 883-44-3
EINECS: 212-931-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR