Isovaleric anhydride 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Isopentanoic anhydride
L04385.06
L04385.14
L04385.06EA
680
CZK
L04385.06
L04385.14
Isovaleric anhydride 95%
Isovaleric anhydride
Purity: 95%
CAS No.: 1468-39-9
Molecular Formula: C10H18O3
Formula Weight: 186.25
Flash Point: 91°(195°F)
Refractive Index: 1.4165
Melting Point: -65°
Boiling Point: 103-104°/24mm
Beilstein Reference: 1635850
UN No.: 3265
MDL No.: MFCD00015051
Vzorec:
C₁₀H₁₈O₃ MW: 186,25 g/mol Bod varu: 103…104 °C (24 mmHg) Bod tání: –65 °C Hustota: 0,927 Bod vzplanutí: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015051 CAS číslo: 1468-39-9 EINECS: 215-994-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...