Tisk…

N,N-Diethyl-1,1-dimethylsilylamine 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: DMSDEA, N,N-Diethyldimethylsilylamine, N-(Dimethylsilyl)diethylamine, N,N-Diethylaminodimethylsilane, Dimethylsilyldiethylamine, (Diethylamino)dimethylsilane

L03503.14 L03503.06
L03503.14EA 6590 CZK
L03503.14 L03503.06
N,N-Diethyl-1,1-dimethylsilylamine 95%
N,N-Diethyl-1,1-dimethylsilylamine

Purity: 95% CAS No.: 13686-66-3 Molecular Formula: C6H17NSi Formula Weight: 131.30 Flash Point: -5°(23°F) Refractive Index: 1.4080 Boiling Point: 109-110° Beilstein Reference: 741862 UN No.: 1993 MDL No.: MFCD00048523

Vzorec: C₆H₁₇NSi
MW: 131.29
Bod varu: 109…110 °C
Hustota: 0.800
Bod vzplanutí: <–5 °C (23 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048523
CAS číslo: 13686-66-3
EINECS: 237-202-5
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR