Tisk…

Vinyltrimethylsilane ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trimethyl(vinyl)silane

L02498.09 L02498.18 L02498.30
L02498.09EA 1540 CZK
L02498.09 L02498.18 L02498.30
Vinyltrimethylsilane ≥97%
Vinyltrimethylsilane

Purity: 97% CAS No.: 754-05-2 Molecular Formula: C5H12Si Formula Weight: 100.24 Flash Point: -24°(-11°F) Refractive Index: 1.3910 Odour: Mild Melting Point: -132° Boiling Point: 54-56° Beilstein Reference: 956572 UN No.: 1993 MDL No.: MFCD00008606

Vzorec: C₅H₁₂Si
Bod varu: 54…56 °C
Bod tání: –132 °C
Hustota: 0,690
Bod vzplanutí: –24 °C (–11 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008606
CAS číslo: 754-05-2
EINECS: 212-042-9
ADR: 3,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR