Tisk…

3-(3-Methoxyphenyl)propionic acid ≥98%

Dodavatel: Thermo Fisher Scientific

L01304.06 L01304.14
L01304.06EA 1520 CZK
L01304.06 L01304.14
3-(3-Methoxyphenyl)propionic acid ≥98%
3-(3-Methoxyphenyl)propionic acid

Purity: 98+% CAS No.: 10516-71-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 43-47° Beilstein Reference: 1874422

Vzorec: C₁₀H₁₂O₃
Bod tání: 43…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00014027
CAS číslo: 10516-71-9
EINECS: 234-049-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR