Tisk…

(3-Methoxyphenoxy)acetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-(3-Methoxyphenoxy)acetic acid

L01100.06 L01100.14
L01100.06EA 1820 CZK
L01100.06 L01100.14
(3-Methoxyphenoxy)acetic acid ≥98%
(3-Methoxyphenoxy)acetic acid

Purity: 98+% CAS No.: 2088-24-6 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 116-119° Beilstein Reference: 1876898

Vzorec: (C₆H₄(OCH₃))OCH₂COOH
Bod tání: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00014357
CAS číslo: 2088-24-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR