(2-Chlorophenoxy)acetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-CPA
L00926.14
L00926.22
L00926.14EA
2310
CZK
L00926.14
L00926.22
(2-Chlorophenoxy)acetic acid ≥98%
(2-Chlorophenoxy)acetic acid
Purity: 98+%
CAS No.: 614-61-9
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
Melting Point: 147-152°
Beilstein Reference: 1103151
Vzorec:
(C₆H₄Cl)OCH₂COOH MW: 186.59 Bod varu: 308 Bod tání: 147…152 °C Hustota: 1.509 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004298 CAS číslo: 614-61-9 EINECS: 210-388-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...