Tetrabromopyrocatechol 96%
Dodavatel: Thermo Scientific
|
Varování
|
L00690.06
L00690.14
L00690.06EA
1060
CZK
L00690.06
L00690.14
Tetrabromopyrocatechol 96%
Tetrabromopyrocatechol
Purity: 96%
CAS No.: 488-47-1
Molecular Formula: C6H2Br4O2
Formula Weight: 425.70
Melting Point: 189-193°
Beilstein Reference: 2051548
Vzorec:
C₆H₂Br₄O₂ Bod varu: 379 °C (1013 hPa) Bod tání: 189…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002189 CAS číslo: 488-47-1 EINECS: 207-678-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...