Triethylsulphonium iodide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L00667.06
L00667.14
L00667.22
L00667.06EA
1440
CZK
L00667.06
L00667.14
L00667.22
Triethylsulphonium iodide ≥97%
Triethylsulphonium iodide
Purity: 97%
CAS No.: 1829-92-1
Molecular Formula: (CH3CH2)3SI
Formula Weight: 246.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 148° dec.
Beilstein Reference: 3911790
MDL No.: MFCD00051867
Vzorec:
C₆H₁₅IS MW: 246.16 Bod tání: ∼ 148 °C (rozkl.) |
MDL Number:
MFCD00051867 CAS číslo: 1829-92-1 EINECS: 217-383-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...