Tisk…

Ethyl phenylpropiolate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Phenylpropiolic acid ethyl ester, Ethyl 3-phenylprop-2-ynoate, Ethyl phenylacetylenecarboxylate

L00603.06 L00603.14
L00603.06EA 1100 CZK
L00603.06 L00603.14
Ethyl phenylpropiolate ≥98%
Ethyl phenylpropiolate

Purity: 98+% CAS No.: 2216-94-6 Molecular Formula: C6H5C≡CCO2CH2CH3 Formula Weight: 174.20 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5520 Odour: Odourless Boiling Point: 152-153°/21mm Beilstein Reference: 639637 MDL No.: MFCD00009185

Vzorec: C₁₁H₁₀O₂
MW: 174,2 g/mol
Bod varu: 152…153 °C (21 mmHg)
Bod tání: 60 °C
Hustota: 1,055
Bod vzplanutí: >110 °C (230 °F)
MDL Number: MFCD00009185
CAS číslo: 2216-94-6
EINECS: 218-703-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR