N-Methylmaleimide ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Methyl-1H-pyrrole-2,5-dione
L00437.06
L00437.14
L00437.06EA
1820
CZK
L00437.06
L00437.14
N-Methylmaleimide ≥98%
N-Methylmaleimide
Purity: 98+%
CAS No.: 930-88-1
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Storage Temperature: 2°C to 8°C
Odour: Odourless
Beilstein Reference: 108550
UN No.: 2928
MDL No.: MFCD00005508
Vzorec:
C₅H₅NO₂ MW: 111,1 g/mol Bod varu: 194 °C (1013 hPa) Bod tání: 94…96 °C |
MDL Number:
MFCD00005508 CAS číslo: 930-88-1 EINECS: 213-226-1 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...