Tisk…

N-ethylmaleimid (NEM) ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

L00355.09 L00355.18
L00355.09EA 3260 CZK
L00355.09 L00355.18
N-ethylmaleimid (NEM) ≥98%
N-ethylmaleimid (NEM)

Purity: 98+% CAS No.: 128-53-0 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Flash Point: 73°(163°F) Odour: Pungent Boiling Point: 209-211° Beilstein Reference: 112448 Merck Reference: 14,3822 UN No.: 2928 MDL No.: MFCD00005509

Vzorec: C₆H₇NO₂
Bod varu: 209…211 °C
Bod tání: 43…46 °C
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005509
CAS číslo: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 14,3822

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR