Tisk…

4-Amino-3-iodopyridine 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Iodopyridin-4-amine, 3-Iodo-4-pyridylamine

H50027.03 H50027.06
H50027.03EA 4430 CZK
H50027.03 H50027.06
4-Amino-3-iodopyridine 97%
4-Amino-3-iodopyridine

Purity: 97% CAS No.: 88511-27-7 Molecular Formula: C5H5IN2 Formula Weight: 220.01 MDL No.: MFCD02663853

Vzorec: C₅H₅IN₂
MW: 220,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663853
CAS číslo: 88511-27-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR