4-Bromo-o-cresol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H32307.06
H32307.14
H32307.06EA
2510
CZK
H32307.06
H32307.14
4-Bromo-o-cresol ≥97%
4-Bromo-o-cresol
Purity: 97%
CAS No.: 2362-12-1
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Odour: Phenol-like
Melting Point: 62-66°
MDL No.: MFCD00055435
Vzorec:
C₇H₇BrO Bod tání: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055435 CAS číslo: 2362-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...