2-Bromo-6-chlorobenzoic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
H32269.03
H32269.06
H32269.03EA
1070
CZK
H32269.03
H32269.06
2-Bromo-6-chlorobenzoic acid ≥98%
2-Bromo-6-chlorobenzoic acid
Purity: 98%
CAS No.: 93224-85-2
Molecular Formula: C7H4BrClO2
Formula Weight: 235.47
Melting Point: 148-150°
MDL No.: MFCD00672929
Vzorec:
C₇H₄BrClO₂ MW: 235,46 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672929 CAS číslo: 93224-85-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...