2-Bromo-6-fluoropyridine ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H30924.03
H30924.06
H30924.03EA
2860
CZK
H30924.03
H30924.06
2-Bromo-6-fluoropyridine ≥97%
2-Bromo-6-fluoropyridine
Purity: 97%
CAS No.: 144100-07-2
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Melting Point: 30-34°
UN No.: 2811
MDL No.: MFCD03095258
Vzorec:
C₅H₃BrFN MW: 175,99 g/mol Bod tání: 30…34 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095258 CAS číslo: 144100-07-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...