4-Ethylbenzyl alcohol ≥99%
Dodavatel: Thermo Scientific
Synonyms:
(4-Ethylphenyl)methanol
H30459.03
H30459.06
H30459.03EA
1360
CZK
H30459.03
H30459.06
4-Ethylbenzyl alcohol ≥99%
4-Ethylbenzyl alcohol
Purity: 99%
CAS No.: 768-59-2
Molecular Formula: C9H12O
Formula Weight: 136.19
Refractive Index: 1.5275
Boiling Point: 115-117°/9mm
MDL No.: MFCD00004666
Vzorec:
C₉H₁₂O MW: 136,19 g/mol Bod varu: 115…117 °C (9 mmHg) Hustota: 1,028 Storage Temperature: Ambient |
MDL Number:
MFCD00004666 CAS číslo: 768-59-2 EINECS: 292-198-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...