Tisk…

4-Chloro-m-anisidine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

H28331.14 H28331.06
H28331.14EA 6830 CZK
H28331.14 H28331.06
4-Chloro-m-anisidine ≥98%
4-Chloro-m-anisidine

Purity: 98% CAS No.: 13726-14-2 Molecular Formula: C7H8ClNO Formula Weight: 157.60 Odour: Characteristic Melting Point: 78-81° UN No.: 2233 MDL No.: MFCD00672967

Vzorec: C₇H₈ClNO
Bod tání: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00672967
CAS číslo: 13726-14-2
UN: 2233
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR