Tisk…

2-Amino-6-chlorobenzothiazole 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Chlorobenzothiazol-2-ylamine, 6-Chloro-1,3-benzothiazol-2-amine

H27877.06 H27877.14
H27877.06EA 1240 CZK
H27877.06 H27877.14
2-Amino-6-chlorobenzothiazole 99%
2-Amino-6-chlorobenzothiazole

Purity: 99% CAS No.: 95-24-9 Molecular Formula: C7H5ClN2S Formula Weight: 184.64 Melting Point: 199-201° MDL No.: MFCD00053557

Vzorec: C₇H₅ClN₂S
MW: 184,65 g/mol
Bod varu: 344 °C (1013 hPa)
Bod tání: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00053557
CAS číslo: 95-24-9
EINECS: 202-402-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR