3-(3-Fluorophenyl)propionic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-(3-Fluorophenyl)propanoic acid
H27847.06
H27847.03
H27847.06EA
2130
CZK
H27847.06
H27847.03
3-(3-Fluorophenyl)propionic acid ≥97%
3-(3-Fluorophenyl)propionic acid
Purity: 97%
CAS No.: 458-45-7
Molecular Formula: C9H9FO2
Formula Weight: 168.17
Flash Point: 110°(230°F)
Melting Point: 43-47°
Boiling Point: 125°/10mm
MDL No.: MFCD01310821
Vzorec:
C₉H₉FO₂ MW: 168.17 Bod varu: 125 °C (10 mmHg) Bod tání: 43…47 °C Bod vzplanutí: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01310821 CAS číslo: 458-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...