Tisk…

3-Bromopyridine-2-carbonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H27632.03 H27632.06
H27632.03EA 2730 CZK
H27632.03 H27632.06
3-Bromopyridine-2-carbonitrile ≥98%
3-Bromopyridine-2-carbonitrile

Purity: 98% CAS No.: 55758-02-6 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Melting Point: 96-99° Boiling Point: 120°/4mm MDL No.: MFCD02683288

Vzorec: C₆H₃BrN₂
Bod varu: 120 °C (4 mmHg)
Bod tání: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD02683288
CAS číslo: 55758-02-6
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR