4-(2-Aminoethyl)pyridine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-(4-Pyridyl)ethylamine, 2-Pyridin-4-yl-ethylamine
H27545.03
H27545.06
H27545.03EA
5560
CZK
H27545.03
H27545.06
4-(2-Aminoethyl)pyridine 97%
4-(2-Aminoethyl)pyridine
Purity: 97%
CAS No.: 13258-63-4
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point:
Vzorec:
C₇H₁₀N₂ MW: 122,17 g/mol Bod varu: 70…71 °C (1 mmHg) Bod tání: 76 °C Hustota: 1,021 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038045 CAS číslo: 13258-63-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...