Tisk…

3-Iodopyridine-2-carbonitrile 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Iodopicolinonitrile, 2-Cyano-3-iodopyridine

H26963.MD H26963.03
H26963.MDEA 2840 CZK
H26963.MD H26963.03
3-Iodopyridine-2-carbonitrile 95%
3-Iodopyridine-2-carbonitrile

Purity: 95% CAS No.: 827616-52-4 Molecular Formula: C6H3IN2 Formula Weight: 230.01 Melting Point: 98-100° UN No.: 3439 MDL No.: MFCD09037451

Vzorec: C₆H₃IN₂
Bod tání: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD09037451
CAS číslo: 827616-52-4
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR