Tisk…

4-Bromo-o-phenylenediamine 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H26929.22 H26929.14 H26929.06
H26929.22EA 15790 CZK
H26929.22 H26929.14 H26929.06
4-Bromo-o-phenylenediamine 97%
4-Bromo-o-phenylenediamine

Purity: 97% CAS No.: 1575-37-7 Molecular Formula: C6H7BrN2 Formula Weight: 187.04 Melting Point: 64-66° UN No.: 2811 MDL No.: MFCD02660622

Vzorec: C₆H₇BrN₂
Bod tání: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD02660622
CAS číslo: 1575-37-7
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR