Tisk…

2,5-Dibromothiazole ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H26909.06 H26909.14
H26909.06EA 4260 CZK
H26909.06 H26909.14
2,5-Dibromothiazole ≥97%
2,5-Dibromothiazole

Purity: 97% CAS No.: 4175-78-4 Molecular Formula: C3HBr2NS Formula Weight: 242.92 Melting Point: 47-49° MDL No.: MFCD00016891

Vzorec: C₃HBr₂NS
MW: 242.92
Bod tání: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00016891
CAS číslo: 4175-78-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR