4-Heptylaniline ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
H26793.06
H26793.14
H26793.06EA
2010
CZK
H26793.06
H26793.14
4-Heptylaniline ≥98%
4-Heptylaniline
Purity: 98%
CAS No.: 37529-27-4
Molecular Formula: C13H21N
Formula Weight: 191.31
Flash Point:
Vzorec:
C₁₃H₂₁N MW: 191,32 g/mol Bod varu: 159 °C (18 mmHg) Hustota: 0,91 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007928 CAS číslo: 37529-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...