4-Fluoro-o-toluonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H26641.06
H26641.14
H26641.06EA
1860
CZK
H26641.06
H26641.14
4-Fluoro-o-toluonitrile ≥97%
4-Fluoro-o-toluonitrile
Purity: 97%
CAS No.: 147754-12-9
Molecular Formula: C8H6FN
Formula Weight: 135.14
Melting Point: 67-71°
UN No.: 3439
MDL No.: MFCD03095106
Vzorec:
C₈H₆FN MW: 135,14 g/mol Bod tání: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095106 CAS číslo: 147754-12-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...