Tisk…

2-Amino-5-phenylpyridine 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 5-Phenylpyridin-2-amine, 5-Phenyl-2-pyridylamine

H26002.MD H26002.03
H26002.MDEA 1760 CZK
H26002.MD H26002.03
2-Amino-5-phenylpyridine 97%
2-Amino-5-phenylpyridine

Purity: 97% CAS No.: 33421-40-8 Molecular Formula: C11H10N2 Formula Weight: 170.21 Melting Point: 133-137° MDL No.: MFCD01692452

Vzorec: C₁₁H₁₀N₂
Bod tání: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD01692452
CAS číslo: 33421-40-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR