Tisk…

5-Phenyl-1H-tetrazole ≥99%

Dodavatel: Thermo Scientific
Varování

B25664.14 B25664.22
B25664.14EA 1670 CZK
B25664.14 B25664.22
5-Phenyl-1H-tetrazole ≥99%
5-Phenyl-1H-tetrazole

Purity: 99% CAS No.: 18039-42-4 Molecular Formula: C7H6N4 Formula Weight: 146.15 Odour: Odourless Melting Point: ca 215° dec. MDL No.: MFCD00022388

Vzorec: C₇H₆N₄
MW: 146,15 g/mol
Bod varu: 331 °C (1013 hPa)
Bod tání: ∼ 215 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00022388
CAS číslo: 18039-42-4
EINECS: 241-950-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR