Tisk…

Trimethylsulphonium bromide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B25098.14 B25098.22 B25098.36
B25098.14EA 2090 CZK
B25098.14 B25098.22 B25098.36
Trimethylsulphonium bromide ≥98%
Trimethylsulphonium bromide

Purity: 98% CAS No.: 3084-53-5 Molecular Formula: (CH3)3SBr Formula Weight: 157.07 Melting Point: 203° subl. MDL No.: MFCD00051367

Vzorec: C₃H₉BrS
Bod tání: 203 °C (subl.)
Hustota: 1,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051367
CAS číslo: 3084-53-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR