Tisk…

5-Amino-4,6-dichloropyrimidine ≥98%

Dodavatel: Thermo Scientific
Varování

B24985.06 B24985.14
B24985.06EA 5260 CZK
B24985.06 B24985.14
5-Amino-4,6-dichloropyrimidine ≥98%
5-Amino-4,6-dichloropyrimidine

Purity: 98+% CAS No.: 5413-85-4 Molecular Formula: C4H3Cl2N3 Formula Weight: 164.00 Odour: Irritating Melting Point: 143-146° Beilstein Reference: 126885 MDL No.: MFCD00006108

Vzorec: C₄H₃Cl₂N₃
MW: 163,99 g/mol
Bod varu: 262 °C (1013 hPa)
Bod tání: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00006108
CAS číslo: 5413-85-4
EINECS: 226-503-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR