4-Bromo-2-chlorobenzenesulphonyl chloride 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-2-chlorobenzene-1-sulphonyl chloride
B24913.06
B24913.14
B24913.22
B24913.06EA
2040
CZK
B24913.06
B24913.14
B24913.22
4-Bromo-2-chlorobenzenesulphonyl chloride 96%
4-Bromo-2-chlorobenzenesulphonyl chloride
Purity: 96%
CAS No.: 351003-52-6
Molecular Formula: C6H3BrCl2O2S
Formula Weight: 289.96
Melting Point: 76-81°
UN No.: 3261
MDL No.: MFCD03094630
Vzorec:
C₆H₃BrCl₂O₂S MW: 289,96 g/mol Bod tání: 76…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094630 CAS číslo: 351003-52-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...