Tisk…

3,4-Dimethylbenzenesulphonyl chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: o-Xylene-4-sulphonyl chloride, 3,4-Dimethylbenzene-1-sulphonyl chloride

B24806.06 B24806.14
B24806.06EA 2060 CZK
B24806.06 B24806.14
3,4-Dimethylbenzenesulphonyl chloride ≥98%
3,4-Dimethylbenzenesulphonyl chloride

Purity: 98% CAS No.: 2905-30-8 Molecular Formula: C8H9ClO2S Formula Weight: 204.68 Odour: Acrid UN No.: 3261 MDL No.: MFCD00625742

Vzorec: C₈H₉ClO₂S
MW: 204,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00625742
CAS číslo: 2905-30-8
EINECS: 210-337-7
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR