Tisk…

Methyl-2,4-dihydroxybenzoate ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,4-Dihydroxybenzoic acid methyl ester, Methyl 2,4-dihydroxybenzoate

B24757.14 B24757.22
B24757.14EA 1220 CZK
B24757.14 B24757.22
Methyl-2,4-dihydroxybenzoate ≥97%
Methyl-2,4-dihydroxybenzoate

Purity: 97% CAS No.: 2150-47-2 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 116-119° MDL No.: MFCD00002276

Vzorec: C₈H₈O₄
Bod varu: 300 °C (1013 hPa)
Bod tání: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00002276
CAS číslo: 2150-47-2
EINECS: 218-428-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR