4,4'-Dichlorobutyrophenone ≥97%
Dodavatel: Thermo Scientific
Synonyms:
4-Chloro-1-(4-chlorophenyl)butan-1-one, γ,4-Dichlorobutyrophenone, ω,4-Dichlorobutyrophenone
B24608.14
B24608.18
B24608.22
B24608.14EA
1730
CZK
B24608.14
B24608.18
B24608.22
4,4'-Dichlorobutyrophenone ≥97%
4,4'-Dichlorobutyrophenone
Purity: 97%
CAS No.: 40877-09-6
Molecular Formula: C10H10Cl2O
Formula Weight: 217.09
Refractive Index: 1.5600
Melting Point: 28-32°
Beilstein Reference: 879782
MDL No.: MFCD00018993
Vzorec:
C₁₀H₁₀Cl₂O MW: 217,09 g/mol Bod varu: 135…140 °C (0,9 mmHg) Bod tání: 28…32 °C Hustota: 1,2575 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00018993 CAS číslo: 40877-09-6 EINECS: 255-123-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...