Tisk…

Methyl dichloroacetate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

B24399.22 B24399.36
B24399.22EA 776 CZK
B24399.22 B24399.36
Methyl dichloroacetate ≥99%
Methyl dichloroacetate

Purity: 99% CAS No.: 116-54-1 Molecular Formula: C3H4Cl2O2 Formula Weight: 142.97 Flash Point: 70°(158°F) Refractive Index: 1.4420 Odour: Pungent Melting Point: -52° Boiling Point: 142-144° UN No.: 2299 MDL No.: MFCD00000843

Vzorec: C₃H₄Cl₂O₂
Bod varu: 142…144 °C
Bod tání: –52 °C
Hustota: 1,380
Bod vzplanutí: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000843
CAS číslo: 116-54-1
EINECS: 204-146-8
UN: 2299
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR