Tisk…

6-Methyluracil ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Methylpyrimidine-2,4(1H,3H)-dione, 4-Methyluracil, 6-Methyl-2,4(1H,3H)-pyrimidinedione

B24191.0E B24191.22 B24191.36
B24191.0EEA 11090 CZK
B24191.0E B24191.22 B24191.36
6-Methyluracil ≥97%
6-Methyluracil

Purity: 97% CAS No.: 626-48-2 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 317-320° Beilstein Reference: 115647 Merck Reference: 14,6133 MDL No.: MFCD00006028

Vzorec: C₅H₆N₂O₂
MW: 126,11 g/mol
Bod varu: 358 °C (1013 hPa)
Bod tání: 317…320 °C
Storage Temperature: Ambient
MDL Number: MFCD00006028
CAS číslo: 626-48-2
EINECS: 210-949-4
Merck Index: 14,6133

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR