4'-Bromo-4-chlorobutyrophenone ≥98%
Dodavatel: Thermo Scientific
Synonyms:
1-(4-Bromophenyl)-4-chloro-1-butanone, 4-Bromo-γ-chlorobutyrophenone, 1-(4-Bromophenyl)-4-chlorobutan-1-one, 4-Bromo-ω-chlorobutyrophenone
B24146.14
B24146.22
B24146.14EA
1860
CZK
B24146.14
B24146.22
4'-Bromo-4-chlorobutyrophenone ≥98%
4'-Bromo-4-chlorobutyrophenone
Purity: 98%
CAS No.: 4559-96-0
Molecular Formula: C10H10BrClO
Formula Weight: 261.55
Flash Point:
Vzorec:
C₁₀H₁₀BrClO MW: 261,55 g/mol Bod varu: 135…139 °C (0,3 mmHg) Bod tání: 35…38 °C Hustota: 1,4808 g/cm³ (20 °C) Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001006 CAS číslo: 4559-96-0 EINECS: 224-930-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...