Tisk…

1-Amino-3-methoxypropane 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Methoxypropylamine

B24095.AK B24095.AU
B24095.AKEA 1100 CZK
B24095.AK B24095.AU
1-Amino-3-methoxypropane 99%
1-Amino-3-methoxypropane

Purity: 99% CAS No.: 5332-73-0 Molecular Formula: CH3O(CH2)3NH2 Formula Weight: 89.14 Flash Point: 27°(81°F) Refractive Index: 1.4175 Odour: Amine-like Melting Point: -70° Boiling Point: 117-119° Beilstein Reference: 878144 UN No.: 2734 MDL No.: MFCD00014831

Vzorec: C₄H₁₁NO
Bod varu: 117…119 °C
Bod tání: < –70 °C
Hustota: 0,872
Bod vzplanutí: 27 °C (81 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014831
CAS číslo: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR