Tisk…

Benzyltributylammonium bromide ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B23941.14 B23941.18 B23941.30
B23941.14EA 1220 CZK
B23941.14 B23941.18 B23941.30
Benzyltributylammonium bromide ≥99%
Benzyltributylammonium bromide

Purity: 99% CAS No.: 25316-59-0 Molecular Formula: [CH3(CH2)3]3N(CH2C6H5)Br Formula Weight: 356.40 Melting Point: ca 175° dec. Beilstein Reference: 3776294

Vzorec: C₁₉H₃₄BrN
Storage Temperature: Ambient
MDL Number: MFCD00011848
CAS číslo: 25316-59-0
EINECS: 246-819-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR