4-Chlorobenzhydrol ≥98%
Dodavatel: Thermo Scientific
Synonyms:
4-Chlorophenyl phenyl carbinol, 4-Chlorobenzhydryl alcohol, 4-Chloro-α-phenylbenzyl alcohol, p-Chlorobenzhydrol, 4-Chlorodiphenylmethanol
B23691.06
B23691.14
B23691.22
B23691.06EA
1250
CZK
B23691.06
B23691.14
B23691.22
4-Chlorobenzhydrol ≥98%
4-Chlorobenzhydrol
Purity: 98%
CAS No.: 119-56-2
Molecular Formula: C13H11ClO
Formula Weight: 218.68
Melting Point: 58-62°
Boiling Point: 150-151°/2.6mm
MDL No.: MFCD00004491
Vzorec:
C₁₃H₁₁ClO MW: 218,68 g/mol Bod varu: 150…151 °C (2,6 mmHg) Bod tání: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004491 CAS číslo: 119-56-2 EINECS: 204-333-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...