2-Amino-6-methylbenzothiazole 99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B23422.14
B23422.22
B23422.14EA
1110
CZK
B23422.14
B23422.22
2-Amino-6-methylbenzothiazole 99%
2-Amino-6-methylbenzothiazole
Purity: 99%
CAS No.: 2536-91-6
Molecular Formula: C8H8N2S
Formula Weight: 164.22
Melting Point: 139-142°
MDL No.: MFCD00005789
Vzorec:
C₈H₈N₂S Bod varu: 321 °C (1013 hPa) Bod tání: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005789 CAS číslo: 2536-91-6 EINECS: 219-801-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...