Tisk…

3-Brompropylamine hydrobromide 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Bromopropan-1-amine hydrobromide, 3-Bromopropylammonium bromide

B23254.14 B23254.22
B23254.14EA 1650 CZK
B23254.14 B23254.22
3-Brompropylamine hydrobromide 98%
3-Brompropylamine hydrobromide

Purity: 98% CAS No.: 5003-71-4 Molecular Formula: Br(CH2)3NH2.HBr Formula Weight: 218.93 Melting Point: 172-174° Beilstein Reference: 3906418

Vzorec: BrH₂CCH₂CH₂NH₃Br
MW: 218.92
Bod tání: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00012912
CAS číslo: 5003-71-4
EINECS: 225-675-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR