Tisk…

3-(Methylthio)aniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-(Methylmercapto)aniline

B23184.06 B23184.14 B23184.22
B23184.06EA 1960 CZK
B23184.06 B23184.14 B23184.22
3-(Methylthio)aniline ≥97%
3-(Methylthio)aniline

Purity: 97% CAS No.: 1783-81-9 Molecular Formula: C7H9NS Formula Weight: 139.23 Flash Point: 110°(230°F) Refractive Index: 1.6370 Odour: Unpleasant Boiling Point: 163-165°/16mm Beilstein Reference: 2078599 UN No.: 2810 MDL No.: MFCD00007793

Vzorec: C₇H₉NS
MW: 139.22
Bod varu: 163…165 °C (16 mmHg)
Hustota: 1.1464
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007793
CAS číslo: 1783-81-9
EINECS: 217-262-5
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR