Tisk…

N,N,N-Trimethylanilinium chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Trimethylphenylammonium chloride, Phenyltrimethylammonium chloride

B22851.22 B22851.36
B22851.22EA 1150 CZK
B22851.22 B22851.36
N,N,N-Trimethylanilinium chloride ≥98%
N,N,N-Trimethylanilinium chloride

Purity: 98+% CAS No.: 138-24-9 Molecular Formula: C6H5N(CH3)3Cl Formula Weight: 171.67 Melting Point: ca 236° subl. Beilstein Reference: 3572527 UN No.: 2811 MDL No.: MFCD00011790

Vzorec: C₉H₁₄ClN
MW: 171,67 g/mol
Bod tání: ∼ 236 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011790
CAS číslo: 138-24-9
EINECS: 205-319-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR