Tisk…

4-(Trifluoromethyl)benzylamine 97%

Dodavatel: Thermo Scientific
Nebezpečí

B22745.03 B22745.06 B22745.14
B22745.03EA 1420 CZK
B22745.03 B22745.06 B22745.14
4-(Trifluoromethyl)benzylamine 97%
4-(Trifluoromethyl)benzylamine

Purity: 98% CAS No.: 3300-51-4 Molecular Formula: C8H8F3N Formula Weight: 175.15 Flash Point: 75°(167°F) Refractive Index: 1.4640 Boiling Point: 80-82°/15mm Beilstein Reference: 2640698 UN No.: 2735 MDL No.: MFCD00010220

Vzorec: C₈H₈F₃N
MW: 175,15 g/mol
Bod varu: 80…82 °C (15 mmHg)
Hustota: 1,229
Bod vzplanutí: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010220
CAS číslo: 3300-51-4
EINECS: 221-971-9
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR