Tisk…

3-Fluoro-N-methylaniline ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

B22565.09 B22565.18
B22565.09EA 3180 CZK
B22565.09 B22565.18
3-Fluoro-N-methylaniline ≥97%
3-Fluoro-N-methylaniline

Purity: 97% CAS No.: 1978-37-6 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 86°(187°F) Odour: Characteristic Boiling Point: 94°/19mm UN No.: 2810 MDL No.: MFCD02683095

Vzorec: C₇H₈FN
MW: 125,15 g/mol
Bod varu: 94 °C (19 mmHg)
Hustota: 1,11
Bod vzplanutí: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD02683095
CAS číslo: 1978-37-6
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR